Learn More
Invitrogen™ Human Carbonic Anhydrase VIII (aa 189-279) Control Fragment Recombinant Protein
Recombinant Protein
Brand:  Invitrogen™ RP93924
Product Code. 30198294
  • 259.00 € / 100µL
Explore more special offers
Description
Description
The protein encoded by this gene was initially named CA-related protein because of sequence similarity to other known carbonic anhydrase genes. However, the gene product lacks carbonic anhydrase activity (i.e., the reversible hydration of carbon dioxide). The gene product continues to carry a carbonic anhydrase designation based on clear sequence identity to other members of the carbonic anhydrase gene family. The absence of CA8 gene transcription in the cerebellum of the lurcher mutant in mice with a neurologic defect suggests an important role for this acatalytic form.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P35219
Blocking Assay, Control
767
100 μL
RUO
Ca8
Human
IQYKGKSKTIPCFNPNTLLPDPLLRDYWVYEGSLTIPPCSEGVTWILFRYPLTISQLQIEEFRRLRTHVKGAELVEGCDGILGDNFRPTQP
Liquid
≥5.0 mg/mL
1M urea, PBS with no preservative; pH 7.4
Carbonic Anhydrase VIII
-20° C, Avoid Freeze/Thaw Cycles
AW546993; Ca8; Cals; Cals1; CAMRQ3; Car8; carbonate dehydratase; carbonic anhydrase 8; carbonic anhydrase VIII; carbonic anhydrase-like sequence; carbonic anhydrase-like sequence 1; carbonic anhydrase-related protein; CA-related protein; CARP; CA-RP; CARP VIII; CA-RP VIII; CA-VIII; hypothetical protein LOC550233; LOW QUALITY PROTEIN: carbonic anhydrase-related protein; MGC120502; MGC99509; wdl; zgc:110118
Unconjugated
Recombinant
E. coli