Learn More
Invitrogen™ Human Carbonic Anhydrase XII (aa 159-278) Control Fragment Recombinant Protein
Recombinant Protein
Brand:  Invitrogen™ RP89981
Product Code. 30196979
  • 259.00 € / 100µL
Explore more special offers
Description
Description
Carbonic Anhydrase XII is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
O43570
Blocking Assay, Control
771
100 μL
RUO
CA12
Human
TASNKSEGLAVLAVLIEMGSFNPSYDKIFSHLQHVKYKGQEAFVPGFNIEELLPERTAEYYRYRGSLTTPPCNPTVLWTVFRNPVQISQEQLLALETALYCTHMDDPSPREMINNFRQVQ
Liquid
≥5.0 mg/mL
1M urea, PBS with no preservative; pH 7.4
Carbonic Anhydrase XII
-20° C, Avoid Freeze/Thaw Cycles
2310047E01Rik; AI314958; CA12; Car12; Carbonate dehydratase XII; carbonic anhydrase 12; Carbonic anhydrase XII; carbonic anyhydrase 12; carbonic dehydratase; CAXII; CA-XII; HsT18816; membrane-bound carbonic anhydrase 12; T18816; tumor antigen HOM-RCC-3.1.3
Unconjugated
Recombinant
E. coli