Learn More
Invitrogen™ Human Carbonic Anhydrase XIV (aa 19-163) Control Fragment Recombinant Protein
Recombinant Protein
Brand:  Invitrogen™ RP90144
Product Code. 30203169
  • 259.00 € / 100µL
Explore more special offers
Description
Description
CA14 belongs a large family of zinc metalloenzymes that catalyze the versible hydration of carbon dioxide. The protein is predicted to be a type I membrane protein and shares highest sequence similarity with the other transmembrane CA isoform, CA XII; however, they have different patterns of tissue-specific expression and thus may play different physiologic roles.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Q9ULX7
Blocking Assay, Control
23632
100 μL
RUO
CA14
Human
GQHWTYEGPHGQDHWPASYPECGNNAQSPIDIQTDSVTFDPDLPALQPHGYDQPGTEPLDLHNNGHTVQLSLPSTLYLGGLPRKYVAAQLHLHWGQKGSPGGSEHQINSEATFAELHIVHYDSDSYDSLSEAAERPQGLAVLGIL
Liquid
≥5.0 mg/mL
1M urea, PBS with no preservative; pH 7.4
Carbonic Anhydrase XIV
-20° C, Avoid Freeze/Thaw Cycles
AW536446; CA XIV; CA14; Car14; Carbonate dehydratase XIV; carbonic anhydrase 14; Carbonic anhydrase XIV; carbonic dehydratase; Catm; CAXiV; CA-XIV; UNQ690/PRO1335
Unconjugated
Recombinant
E. coli