Learn More
Invitrogen™ Human Caspase-1 ELISA Kit
Human Caspase-1 quantitates human Caspase-1 in serum, plasma, supernatant.
Brand:  Invitrogen™ EH70RBX5
 View more versions of this product
Product Code. 17195299
  • 2690.00 € / Each
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Human Caspase-1 quantitates human Caspase-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Caspase-1.
Caspases are a family of cysteine proteases that can be divided into the apoptotic and inflammatory caspase subfamilies. Unlike the apoptotic caspases, members of the inflammatory subfamily are generally not involved in cell death but are associated with the immune response to microbial pathogens. Members of this subfamily include caspase-1, -4, -5, and -12 and can activate proinflammatory cytokines such as IL-1-b and IL-18. Caspase-1 was initially identified as an IL-1-b-converting enzyme; later experiments revealed it to be a mammalian homolog of the C. elegans cell death gene ced-3 whose overexpression can induce apoptosis in fibroblasts.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CASP1; CASP-1; CASP1 nirs variant 1; caspase 1; caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, convertase); Caspase1; caspase-1; Caspase-1 subunit p10; Caspase-1 subunit p20; ICE; IL-1 beta-converting enzyme; IL-1 B converting enzyme; IL1BC; IL-1 BC; IL1BCE; IL1B-convertase; interleukin 1 beta-converting enzyme; interleukin 1, beta, convertase; interleukin 1-B converting enzyme; Interleukin 1 beta converting enzyme; interleukin-1 beta convertase; interleukin-1 beta-converting enzyme; p45
0.43 ng/mL
ELISA
Colorimetric Microplate Reader
CASP1
<10%
HRP
Research Use Only
2°C to 8°C
1 hr 20 min
0.43 to 100 ng/mL
Biotin
Plasma, Serum, Supernatant
P29466
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
5 x 96 Tests
Serum, 50 μL; Plasma, 50 μL; Supernatant, 50 μL
Caspase 1
4 hr 45 min