Learn More
Invitrogen™ Human Caspase 6 Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP0234
Product Code. 15276449
  • 141.00 € / 50µg
Explore more special offers
Description
Description
Caspase-6 Synthetic Peptide, PEP-0234, from Invitrogen.
Caspases are cysteine proteases, expressed as inactive precursors, that mediate apoptosis by proteolysis of specific substrates. Caspases have the ability to cleave after aspartic acid residues. There are two classes of caspases involved in apoptosis; initiators (activation by receptor cluster) and effectors (activation by mitochondrial permeability transition). Proapoptotic signals autocatalytically activate initiator caspases, such as Caspase-8 and Caspase 9. Activated initiator caspases then process effector caspases, such as Caspase-3 and Caspase-7, which in turn cause cell collapse.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
839
purified
Human Caspase 6
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
P55212
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
CASP6
0.02% sodium azide
50 μg
PBS
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Human Caspase 6 Synthetic Peptide > 50 μg; Unlabeled