Learn More
R&D Systems™ Human CMTM3 (NP_653202.1) VersaClone cDNA
Brand: R&D Systems™ RDC3576
This item is not returnable.
View return policy
Description
Specifications
CMTM3 | |
123920 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
NP_653202.1 | |
BNAS2, chemokine-like factor superfamily 3, Chemokine-like factor superfamily member 3, CKLF-like MARVEL transmembrane domain containing 3, CKLF-like MARVEL transmembrane domain-containing protein 3, CKLFSF3, FLJ31762, MGC51956 | |
Human |