Learn More
Novus Biologicals™ Human Colon Tissue MicroArray (Tumor)
Tissue arrays are 4 - 6 um in thickness, and mounted on positively charged glass slides
Brand:  Novus Biologicals™ NBP2-42062
Product Code. 18618506
  • 692.00 € / Pack of 5
Explore more special offers
Specifications
5 Slides
Human
Immunohistochemistry, Immunohistochemistry-Paraffin