missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CTPS2 (aa 252-302) Control Fragment Recombinant Protein

Product Code. 30195227
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195227

Brand: Invitrogen™ RP109515

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene catalyzes the formation of CTP from UTP with the concomitant deamination of glutamine to glutamate. This protein is the rate-limiting enzyme in the synthesis of cytosine nucleotides, which play an important role in various metabolic processes and provide the precursors necessary for the synthesis of RNA and DNA. Cancer cells that exhibit increased cell proliferation also exhibit an increased activity of this encoded protein. Thus, this protein is an attractive target for selective chemotherapy. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NRF8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 56474
Name Human CTPS2 (aa 252-302) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias A830031M15Rik; AI326475; CTP synthase 2; CTP synthase II; CTP synthetase 2; CTP synthetase homolog; CTP synthetase type 2; CTPS2; CTPsH; cytidine 5'-triphosphate synthase 2; cytidine 5'-triphosphate synthetase 2; cytidine triphosphate synthase II; UTP-ammonia ligase 2; UTP--ammonia ligase 2
Common Name CTPS2
Gene Symbol Ctps2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RVPVLLEEQSIVKYFKERLHLPIGDSASNLLFKWRNMADRYERLQKICSIA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.