missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human DPCR-1 (Q3MIW9) VersaClone cDNA
Diffuse Panbronchiolitis Critical Region 1
Brand: R&D Systems™ RDC1943
This item is not returnable.
View return policy
Description
The Human DPCR-1 (Q3MIW9) VersaClone cDNA clone contains the complete ORF for DPCR-1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The DPCR-1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
cDNA | |
Q3MIW9 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
135656 (Human); 268949 (Mouse); 406199 (Rat) | |
Human |
hDPCR1 | |
10 μg at 0.2 μg/μL | |
1567bp | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction