Learn More
Invitrogen™ Human ECM-1 ELISA Kit
Human ECM-1 quantitates human ECM-1 in serum, plasma, supernatant.
Brand:  Invitrogen™ EH165RB
 View more versions of this product
Product Code. 17174443
  • 656.00 € / Pack of 96
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Human ECM-1 quantitates human ECM-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ECM-1.
Secretory Component is an epithelial transport receptor for polymeric IgA and IgM. Antibody to secretory component has been shown to be useful for identifying glandular carcinomas.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AI663821; ECM1; extracellular matrix protein 1; p85; RP11-54A4.6; Secretory component p85; testicular tissue protein Li 61; URBWD
24 pg/mL
ELISA
Colorimetric Microplate Reader
ECM1
<10%
HRP
Research Use Only
2°C to 8°C
1 hr 20 min
24 to 6,000 pg/mL
Biotin
Plasma, Serum, Supernatant
Q16610
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL
ECM1
4 hr 45 min