missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human EGF Recombinant Protein

Human EGF Recombinant Protein

Brand:  Gibco™ PHG0311L

Product Code. 10125414

  • 275.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Human EGF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

EGF has a profound effect on the differentiation of specific cells in vivo and is a potent mitogenic factor for a variety of cultured cells of both ectodermal and mesodermal origin.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High-purity, recombinant, bioactive protein: to help ensure growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High biological activity: in-house activity testing of recombinant proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of mouse BALB/3T3 cells

Shipping Condition: Dry Ice.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

NM_001963
EGF
Sequential Chromatography
EGF
Human
Store at ≤ 20°C. Upon initial thawing, we recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. The contents should then be apportioned into working aliquots and stored at ≤ 20°C. Avoid repeated freeze⁄thaw cycles. Further dilutions should be made in low endotoxin medium or buffered solution with FBS or tissue culture grade BSA.
< 0.1 ng/μg
100 μg
Dry Ice
E. coli
Recombinant, Ligand
EGF (Epidermal Growth Factor)
Gibco™
Cell Culture
6.2 kDa
100 ug
ED50 range = 0.1-0.4 ng⁄mL, ED50 range = 0.1-0.4 ng⁄mL
Untagged
Carrier-Free
Balb/3T3 cells
Human
1.0 x 107 - 2.5 x 106 units⁄mg
Growth Factors & Receptors
Liquid
Stem Cell Research, Oncology
95 %
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.