Learn More
Novus Biologicals™ Human Esophagus Whole Tissue Lysate (Adult Whole Diabetes)
Brand:  Novus Biologicals™ NB820-59287
Product Code. 18223955
  • 688.00 € / 1mg
Explore more special offers
Description
Description
The Human Esophagus Whole Tissue Lysate (Adult Whole Diabetes) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
Western Blot
1 mg
Total Protein - Human Diabetic Diseased Tissue: Esophagus
Diabetes
Human
Human Esophagus Whole Tissue Lysate (Adult Diabetes)
Adult
Digestive/Waste
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Whole
Human
5 mg/mL
-80°C. Avoid freeze-thaw cycles.
Native