missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Eye Membrane Tissue Lysate (Adult Membrane Normal)

Product Code. 18299243 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
This item is not returnable. View return policy

Product Code. 18299243

Brand: Novus Biologicals™ NB82060632

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The Human Eye Membrane Tissue Lysate (Adult Membrane Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot
Tissue Visual
Quantity 0.1 mg
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Membrane Protein - Human Adult Normal Tissue: Whole Eye
Cellular Fraction Membrane
Tissue Condition Normal
Cross Reactivity Human
Host Species Human
Concentration 2 mg/mL
Product Type Human Eye Membrane Tissue Lysate (Adult Normal)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.