Learn More
Gibco™ Human FGF-17 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand:  Gibco™ 100-27-500
 View more versions of this product
Product Code. 30089027
  • 1818.00 € / Pack of 2
Explore more special offers
Description
Description
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes including embryonic development cell growth, morphogenesis, tissue repair, tumor growth and invasion. This gene was shown to be prominently expressed in the cerebellum and cortex. The mouse homolog of this gene was localized to specific sites in the midline structures of the forebrain, the midbrain-hindbrain junction, developing skeleton and developing arteries, which suggests a role in central nervous system, bone and vascular development. This gene was referred to as FGF-13 in reference 2, however, its amino acid sequence and chromosomal localization are identical to FGF17.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
O60258
Protein with no preservative
22.7
2 x 250 μg
-20°C
<1 EU/μg
FGF17
Unconjugated
MTQGENHPSP NFNQYVRDQG AMTDQLSRRQ IREYQLYSRT SGKHVQVTGR RISATAEDGN KFAKLIVETD TFGSRVRIKG AESEKYICMN KRGKLIGKPS GKSKDCVFTE IVLENNYTAF QNARHEGWFM AFTRQGRPRQ ASRSRQNQRE AHFIKRLYQG QLPFPNHAEK QKQFEFVGSA PTRRTKRTRR PQPLT
≥ 95% by SDS-PAGE gel and HPLC analyses.
ELISA, Western Blot, Functional Assay
8822
Human FGF-17
E. coli
RUO
FGF; FGF-13; FGF17; FGF-17; Fibroblast growth factor; Fibroblast growth factor 17; HH20; UNQ161/PRO187
Human
Recombinant
Lyophilized