missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-23 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
80.50 € - 3865.00 €
Specifications
Accession Number | Q9GZV9 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 8074 |
Molecular Weight (g/mol) | 25.5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17841253
|
Gibco™
100-52-100UG |
100 μg |
772.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17801513
|
Gibco™
100-52-1MG |
1 mg |
3865.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17841143
|
Gibco™
100-52-20UG |
20 μg |
195.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17811343
|
Gibco™
100-52-250UG |
250 μg |
1356.00 €
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17830273
|
Gibco™
100-52-500UG |
500 μg |
2325.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17870853
|
Gibco™
100-52-5UG |
5 μg |
80.50 €
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The product of this gene inhibits renal tubular phosphate transport. This gene was identified by its mutations associated with autosomal dominant hypophosphatemic rickets, an inherited phosphate wasting disorder. Abnormally high level expression of this gene was found in oncogenic hypophosphatemic osteomalacia, a phenotypically similar disease caused by abnormal phosphate metabolism. Mutations in this gene have also been shown to cause familial tumoral calcinosis with hyperphosphatemia.Specifications
Q9GZV9 | |
protein with no preservative | |
25.5 | |
E. coli | |
RUO | |
ADHR; FGF; Fgf23; FGF-23; FGFN; Fibroblast growth factor; Fibroblast growth factor 23; Fibroblast growth factor 23 C-terminal peptide; Fibroblast growth factor 23 N-terminal peptide; HPDR2; HYPF; Phosphatonin; PHPTC; tumor-derived hypophosphatemia inducing factor; Tumor-derived hypophosphatemia-inducing factor; UNQ3027/PRO9828 | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
8074 | |
Human FGF-23 | |
-20°C | |
<1 EU/μg | |
FGF23 | |
Unconjugated | |
MYPNASPLLG SSWGGLIHLY TATARNSYHL QIHKNGHVDG APHQTIYSAL MIRSEDAGFV VITGVMSRRY LCMDFRGNIF GSHYFDPENC RFQHQTLENG YDVYHSPQYH FLVSLGRAKR AFLPGMNPPP YSQFLSRRNE IPLIHFNTPI PRRHTRSAED DSERDPLNVL KPRARMTPAP ASCSQELPSA EDNSPMASDP LGVVRGGRVN THAGGTGPEG CRPFAKFI | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |