missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-BP-1 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 100-66-500UG
This item is not returnable.
View return policy
Description
FGFBP1 encodes a secreted fibroblast growth factor carrier protein. The encoded protein plays a critical role in cell proliferation, differentiation and migration by binding to fibroblast growth factors and potentiating their biological effects on target cells. The encoded protein may also play a role in tumor growth as an angiogenic switch molecule, and expression of this gene has been associated with several types of cancer including pancreatic and colorectal adenocarcinoma. A pseudogene of this gene is also located on the short arm of chromosome 4.Specifications
Q14512 | |
protein with no preservative | |
23.9 | |
500 μg | |
-20°C | |
<1 EU/μg | |
FGFBP1 | |
Unconjugated | |
MKKKVKNGLH SKVVSEQKDT LGNTQIKQKS RPGNKGKFVT KDQANCRWAA TEQEEGISLK VECTQLDHEF SCVFAGNPTS CLKLKDERVY WKQVARNLRS QKDICRYSKT AVKTRVCRKD FPESSLKLVS STLFGNTKPR KEKTEMSPRE HIKGKETTPS SLAVTQTMAT KAPECVEDPD MANQRKTALE FCGETWSSLC TFFLSIVQDT SC | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
9982 | |
Human FGF-BP-1 | |
E. coli | |
RUO | |
17 kDa HBGF-binding protein; 17 kDa heparin-binding growth factor-binding protein; FGF-binding protein 1; FGFBP; FGF-BP; Fgfbp1; FGF-BP1; FGFBP-1; fibroblast growth factor binding protein 1; fibroblast growth factor-binding protein 1; growth factor binding protein-1; growth factor-binding protein 1; HBP17; heparin-binding growth factor binding protein | |
Human | |
Recombinant | |
Lyophilized |