missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGFR2a (IIIc) Fc Recombinant Protein, PeproTech®
CHO cells Recombinant Protein
Brand: Gibco™ 160-03-50UG
130.00 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
The FGF family plays a central role during prenatal development and postnatal growth, and the regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. The FGF ligands bind to a family of type I transmembrane tyrosine kinase receptors, which leads to dimerization and activation by sequential autophosphorylation of specific tyrosine residues. Four genes encoding structurally related FGF receptors (FGFR-1 to -4) are known. Alternative splicing of the mRNAs generates numerous forms of FGFR-1 to -3. Alternate forms of FGF receptors can exhibit different specificities with respect to ligand binding. For example, the form designated as FGFR1a (IIc) interacts predominantly with FGF-acidic (FGF1) and FGF-basic (FGF2). A frequent splicing event involving FGFR-1 and -2 results in receptors containing all three Ig domains, referred to as the alpha isoform, or only IgII and IgIII, referred to as thebeta isoform. Only the alpha isoform has been identified for FGFR-3 and FGFR-4. Additional splicing events for FGFR-1 to -3, involving the C-terminal half of the IgIII domain encoded by two mutually exclusive alternative exons, generate FGF receptors with alternative IgIII domains (IIIb and IIIc).Specifications
P21802 | |
protein with no preservative | |
65.6 | |
50 μg | |
-20°C | |
<1 EU/μg | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
2263 | |
Human FGFR2a (IIIc) Fc | |
CHO cells | |
RUO | |
FGFR2 | |
Unconjugated | |
RPSFSLVEDT TLEPEEPPTK YQISQPEVYV AAPGESLEVR CLLKDAAVIS WTKDGVHLGP NNRTVLIGEY LQIKGATPRD SGLYACTASR TVDSETWYFM VNVTDAISSG DDEDDTDGAE DFVSENSNNK RAPYWTNTEK MEKRLHAVPA ANTVKFRCPA GGNPMPTMRW LKNGKEFKQE HRIGGYKVRN QHWSLIMESV VPSDKGNYTC VVENEYGSIN HTYHLDVVER SPHRPILQAG LPANASTVVG GDVEFVCKVY SDAQPHIQWI KHVEKNGSKY GPDGLPYLKV LKAAGVNTTD KEIEVLYIRN VTFEDAGEYT CLAGNSIGIS FHSAWLTVLP APGREKEITA SPDYLEGGPK SCDKTHTCPP CPAPELLGGP SVFLFPPKPK DTLMISRTPE VTCVVVDVSH EDPEVKFNWY VDGVEVHNAK TKPREEQYNS TYRVVSVLTV LHQDWLNGKE YKCKVSNKAL PAPIEKTISK AKGQPREPQV YTLPPSRDEL TKNQVSLTCL VKGFYPSDIA VEWESNGQPE NNYKTTPPVL DSDGSFFLYS KLTVDKSRWQ QGNVFSCSVM HEALHNHYTQ KSLSLSPGK | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |