missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human GDNF Recombinant Protein
Glial-Derived Neurotrophic Factor (GDNF) is a bioactive protein intended for use in cell culture applications
416.00 € - 6850.00 €
Specifications
For Use With (Application) | Cell Culture |
---|---|
Molecular Weight (g/mol) | 30.1 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | G-DNF |
Species | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10110633
|
Gibco™
PHC7044 |
5 μg |
416.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10679963
|
Gibco™
PHC7045 |
10 μg |
558.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10641504
|
Gibco™
PHC7041 |
100 μg |
1946.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10737683
|
Gibco™
PHC7043 |
1 mg |
6850.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Human GDNF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
GDNF is mainly known for its affect on neurons, but it is also involved in other biological processes including kidney development.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by inducing neurite outgrowth on PC-12 cells in the presence of recombinant soluble GFR-α
- Proven Compatibility: Gibco proteins are bioassayed with Gibco media
Specifications
Cell Culture | |
Sequential Chromatography | |
Human | |
Approved for shipment on Wet or Dry Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Neurobiology, Stem Cell Research, Differentiation | |
98 % |
30.1 kDa | |
G-DNF | |
Carrier-Free | |
E. coli | |
Recombinant, Ligand | |
Neurotrophic Factors | |
Gibco™ |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.