missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human GLP-1 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
80.50 € - 692.00 €
Specifications
Accession Number | P01275 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 2641 |
Molecular Weight (g/mol) | 3.3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17861643
|
Gibco™
130-08-1MG |
1 mg |
195.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17841743
|
Gibco™
130-08-200UG |
200 μg |
80.50 €
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17871443
|
Gibco™
130-08-5MG |
5 mg |
692.00 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GCG is also known as GLP1, or Glucagon. Glucagon is a 29-amino acid pancreatic hormone that counteracts the glucose-lowering action of insulin by stimulating glycogenolysis and gluconeogenesis.It is mapped to 2q36-2q37. GLP1, also known as 7-37 for the codons of the preproglucagon molecule which encode it, renders pancreatic beta-cells 'glucose-competent' and may be useful in the treatment of noninsulin-dependent diabetes mellitus. Also, GLP1 is a potent insulin secretagogue.It plays a major role in the enteroinsular axis, accounting, for example, for the finding that plasma insulin levels accompanying oral intake of glucose are greater than those observed when glucose is given intravenously.Specifications
P01275 | |
protein with no preservative | |
3.3 | |
E. coli | |
RUO | |
GCG; Glicentin; Glicentin-related polypeptide; GLP1; GLP-1; GLP-1(7-36); GLP-1(7-37); GLP1/2; GLP2; GLP-2; Glu; glucagon; Glucagon like peptide 1; Glucagon-like peptide 1; Glucagon-like peptide 1(7-36); Glucagon-like peptide 1(7-37); Glucagon-like peptide 2; glucagon-like peptide-1; GRPP; Incretin hormone; MGC138331; OXM; OXY; Oxyntomodulin; PPG; preproglucagon | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
2641 | |
Human GLP-1 | |
-20°C | |
<1 EU/μg | |
GCG | |
Unconjugated | |
HAEGTFTSDV SSYLEGQAAK EFIAWLVKGR G | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title