Learn More
Invitrogen™ Human Glucocorticoid Receptor beta (aa 728-742) Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP222
Product Code. 11534870
  • 158.00 € / 50µg
Explore more special offers
Description
Description
Human GR beta (residues 728-742) Synthetic Peptide, PEP-222, from Invitrogen.
PEP-222 is a 16 amino acid synthetic peptide corresponding to amino acid residues 728-742 from human GR beta. The sequence of this peptide is (amino to carboxy terminus): N (728) V M W L K P E S T S H T L I (742) CThis peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, catalog # PA3-514. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2908
purified
Human Glucocorticoid Receptor beta (aa 728-742)
-20°C. Avoid freeze-thaw cycles.
Lyophilized
Control
Yes
Peptide
RUO - research use only
Human
P04150
0.5 mg/mL
Synthetic Peptide
peptide with no preservative
NR3C1
No preservative
50 μg
N(728) V M W L K P E S T S H T L I(742) C