missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human GNMT ELISA Kit
Human GNMT quantitates human GNMT in serum, plasma, supernatant.
Brand: Invitrogen™ EH226RBX10
This item is not returnable.
View return policy
Description
Human GNMT quantitates human GNMT in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human GNMT.
The protein encoded by this gene is an enzyme that catalyzes the conversion of S-adenosyl-L-methionine to S-adenosyl-L-homocysteine and sarcosine. The encoded protein is found in the cytoplasm and acts as a homotetramer. Defects in this gene are a cause of GNMT deficiency.Specifications
2.1.1.20; epididymis secretory sperm binding protein Li 182 mP; folate-binding protein; glycine methyltransferase; glycine N methyl transferase; glycine N-methyltransferase; GNMT; HEL-S-182 mP | |
2.5 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
GNMT | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
2.5 to 500 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Q14749 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Supernatant, 50 μL; Plasma, 50 μL; Serum, 50 μL | |
GNMT | |
4 hr 45 min |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction