Learn More
Invitrogen™ Human Hemopexin ELISA Kit
Human Hemopexin quantitates human Hemopexin in serum, plasma, supernatant.
Brand:  Invitrogen™ EH238RBX5
 View more versions of this product
Product Code. 17134999
  • 2690.00 € / Each
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Human Hemopexin quantitates human Hemopexin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Hemopexin.
Hemopexin is a serum glycoprotein that binds heme and transports it to the liver for breakdown and iron recovery, after which the free hemopexin returns to the circulation. Structurally, hemopexin consists of two similar halves of approximately two hundred amino acid residues connected by a histidine-rich hinge region. Each half is itself formed by the repetition of a basic unit of some 35 to 45 residues.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Beta-1 B-glycoprotein; hemopexin; Hpx; Hpxn; HX; Hyaluronidase
0.2 ng/mL
ELISA
Colorimetric Microplate Reader
Hpx
<10%
HRP
Research Use Only
2°C to 8°C
1 hr 20 min
0.2 to 50 ng/mL
Biotin
Plasma, Serum, Supernatant
P02790
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
5 x 96 Tests
Supernatant, 50 μL; Plasma, 50 μL; Serum, 50 μL
Hemopexin
4 hr 45 min