missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Hemopexin ELISA Kit
Human Hemopexin quantitates human Hemopexin in serum, plasma, supernatant.
Brand: Invitrogen™ EH238RB
This item is not returnable.
View return policy
Description
Human Hemopexin quantitates human Hemopexin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Hemopexin.
Hemopexin is a serum glycoprotein that binds heme and transports it to the liver for breakdown and iron recovery, after which the free hemopexin returns to the circulation. Structurally, hemopexin consists of two similar halves of approximately two hundred amino acid residues connected by a histidine-rich hinge region. Each half is itself formed by the repetition of a basic unit of some 35 to 45 residues.Specifications
Beta-1 B-glycoprotein; hemopexin; Hpx; Hpxn; HX; Hyaluronidase | |
0.2 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Hpx | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
0.2 to 50 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
P02790 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Supernatant, 50 μL; Plasma, 50 μL; Serum, 50 μL | |
Hemopexin | |
4 hr 45 min |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction