Learn More
R&D Systems™ Human HIF-3 alpha/HIF3A (NP_690008.2) VersaClone cDNA
Contains gene inserts that are fully sequenced and produced with high quality
Brand:  R&D Systems™ RDC3435
Product Code. 18492350
  • 719.00 € / 10µg
Explore more special offers
Specifications
HIF-3 alpha
64344
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
NP_690008.2
Basic-helix-loop-helix-PAS protein MOP7, BHLHE17, bHLHe17HIF-3A2, Class E basic helix-loop-helix protein 17, HIF3A, HIF-3A4, HIF-3-alpha, HIF3-alpha, hypoxia inducible factor 3, alpha subunit, hypoxia-inducible factor 3-alpha, hypoxia-inducible factor-3 alpha 4, Inhibitory PAS domain protein, IPAS, IPASHIF3-alpha-1, Member of PAS protein 7, MOP7, MOP7PAS domain-containing protein 7, PASD7, PASD7HIF-3A