missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human HVEM Fc Recombinant Protein, PeproTech®
Insect cells Recombinant Protein
80.50 € - 1098.00 €
Specifications
Accession Number | Q92956 |
---|---|
For Use With (Application) | Functional Assay |
Gene ID (Entrez) | 8764 |
Molecular Weight (g/mol) | 41.4 |
Name | Human HVEM Fc |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17842293
|
Gibco™
310-27-100UG |
100 μg |
195.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17832563
|
Gibco™
310-27-1MG |
10 x 100 μg | N/A | ||||||
30089289
|
Gibco™
310-27-200 |
2 x 100 μg |
324.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17832363
|
Gibco™
310-27-200UG |
2 x 100 μg | N/A | ||||||
30087939
|
Gibco™
310-27-500 |
5 x 100 μg |
654.00 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17872993
|
Gibco™
310-27-20UG |
20 μg |
80.50 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17842363
|
Gibco™
310-27-500UG |
5 x 100 μg | N/A | ||||||
Description
TNFRSF14 is a member of the TNF-receptor superfamily. TNFRSF14 was identified as a cellular mediator of herpes simplex virus (HSV) entry. Binding of HSV viral envelope glycoprotein D (gD) to TNFRSF14 has been shown to be part of the viral entry mechanism. The cytoplasmic region of TNFRSF14 was found to bind to several TRAF family members, which may mediate the signal transduction pathways that activate the immune response. Activation of the signal transduction pathway involving TNFRSF14 in T cells stimulates T cell proliferation and cytokine production, leading to inflammation and enhanced CTL-mediated tumor immunity, suggesting that these proteins may be useful as potential targets for controlling cellular immune responses. Multiple alternatively spliced transcript variants have been described, but the full-length nature of some of these TNFRSF14 variants have not been determined.Specifications
Q92956 | |
8764 | |
Human HVEM Fc | |
-20°C | |
<1 EU/μg | |
TNFRSF14 | |
Unconjugated | |
LPSCKEDEYP VGSECCPKCS PGYRVKEACG ELTGTVCEPC PPGTYIAHLN GLSKCLQCQM CDPAMGLRAS RNCSRTENAV CGCSPGHFCI VQDGDHCAAC RAYATSSPGQ RVQKGGTESQ DTLCQNCPPG TFSPNGTLEE CQHQTKRSCD KTHTCPPCPA PELLGGPSVF LFPPKPKDTL MISRTPEVTC VVVDVSHEDP EVKFNWYVDG VEVHNAKTKP REEQYNSTYR VVSVLTVLHQ DWLNGKEYKC KVSNKALPAP IEKTISKAKG QPREPQVYTL PPSRDELTKN QVSLTCLVKG FYPSDIAVEW ESNGQPENNY KTTPPVLDSD GSFFLYSKLT VDKSRWQQGN VFSCSVMHEA LHNHYTQKSL SLSPGK | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
41.4 | |
Insect cells | |
RUO | |
Atar; CD270; CD40-like protein; herpes virus entry mediator; herpes virus entry mediator A; Herpesvirus entry mediator A; HGNC:11912; HVEA; Hvem; LIGHTR; RP3-395M20.6; sCD2710; TNF receptor superfamily member 14; Tnfrs14; TNFRSF14; TR2; tumo; Tumor necrosis factor receptor superfamily member 14; tumor necrosis factor receptor superfamily, member 14; tumor necrosis factor receptor superfamily, member 14 (herpesvirus entry mediator); Tumor necrosis factor receptor-like 2; tumor necrosis factor receptor-like gene2; UNQ329/PRO509 | |
Human | |
Recombinant | |
Lyophilized |