Learn More
Invitrogen™ Interferon alpha (Cell culture) Human ELISA Kit
Measure levels of human interferon alpha (IFNa) in human serum and human plasma samples using 96-well plates.
918.00 € - 3595.00 €
Specifications
Assay Range | 12.5-1000pg/mL |
---|---|
Description | Human IFN alpha ELISA Kit (Serum Samples) |
Conjugate | HRP |
Immunoassay Kit Format | Colorimetric ELISA (TMB substrate) with Plates |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10531594
|
Invitrogen™
411101 |
96 Tests |
918.00 €
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10177923
|
Invitrogen™
411102 |
5 x 96 Tests |
3595.00 €
1 set |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
Description
Assay IFN-a (human interferon alpha) levels in human serum, plasma and tissue culture media with these Thermo Scientific™ Human IFN alpha ELISA Kits (for Serum Samples).
These human IFN alpha ELISA kits use pre-titered, matched pairs of coating and detection antibodies to accurately measure interferon alpha (IFN-α) in samples at 50μL per well. These IFN-α kits are validated for use with human serum, plasma and tissue culture media. Also included are calibrated recombinant protein standard, TMB substrate solution and stop solution.The complete kits contain pre-coated 96-well plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.Target: Human interferon alpha (human IFN-α, IFN-alpha)
Format: Colorimetric ELISA (TMB substrate) in clear 96-well plate
Measurement: Absorbance at 450nm
Assay Range: 12.5 to 1000pg/mL
Sensitivity: Undetermined
Precision: Intra-assay CV ≤ 8%; inter-assay CV ≤ 8%
Sample Types: Cell culture supernatant, serum, plasma
Total Assay Time: 3.25 hours
Sample Size: 50μL
Specificity: No significant cross-reaction with human IFN-β, IFN-γ or IFN-ω; mouse or rat IFN-α, IFN-β, or IFN-γ; bovine IFN-τ
Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, substrates, and stop solution.
Specifications
12.5-1000pg/mL | |
HRP | |
ELISA Kit | |
No significant cross-reaction with human IFN-ß, IFN-g or IFN-w; mouse or rat IFN-a, IFN-ß, or IFN-g; bovine IFN-t | |
Colorimetric Microplate Reader | |
<8% | |
HRP | |
Intra-assay CV ≤ 8%; inter-assay CV ≤ 8% | |
Supernatant, 100 μL | |
Human | |
3 hr. 15 min. |
Human IFN alpha ELISA Kit (Serum Samples) | |
Colorimetric ELISA (TMB substrate) with Plates | |
Supernatant | |
Human | |
IFNA1,IFNA2 | |
<8% | |
Absorbance at 450nm | |
RUO | |
2°C to 8°C | |
1 hr. 20 min. |