missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-10 Recombinant Protein, PeproTech®

E. coli Recombinant Protein

80.00 € - 5140.00 €

Specifications

Accession Number P22301
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 3586
Molecular Weight (g/mol) 18.6
View More Specs

Products 7
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17830513
View Documents
Gibco™
200-10-100UG
100 μg
1030.00 €
100µg
Please to purchase this item. Need a web account? Register with us today!
17890563
View Documents
Gibco™
200-10-10UG
10 μg
194.00 €
10µg
Please to purchase this item. Need a web account? Register with us today!
17821303
View Documents
Gibco™
200-10-1MG
1 mg
5140.00 €
1mg
Please to purchase this item. Need a web account? Register with us today!
17880333
View Documents
Gibco™
200-10-250UG
250 μg
1802.00 €
250µg
Please to purchase this item. Need a web account? Register with us today!
17861073
View Documents
Gibco™
200-10-2UG
2 μg
80.00 €
2µg
Please to purchase this item. Need a web account? Register with us today!
17800523
View Documents
Gibco™
200-10-500UG
500 μg
3085.00 €
500µg
Please to purchase this item. Need a web account? Register with us today!
17891073
View Documents
Gibco™
200-10-50UG
50 μg
620.00 €
50µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Interleukin 10 (IL-10, CSIF) is an anti-inflammatory cytokine mainly produced by macrophages and Th2 cells. IL-10 is an 18.6 kDa protein of 160 amino acid residues that shares over 80% sequence homology with the Epstein-Barr Virus protein (BCRFI). The reported biological activities of IL-10, which maybe interrelated, include inhibition of macrophage-mediated cytokine synthesis, suppression of the delayed-type hyper-sensitivity response, and stimulation of the Th2 cell response which results in elevated antibody production. IL-10 functions by inhibiting pro-inflammatory cytokines made by macrophages and regulatory T cells including IFN-gamma, TNF-alpha, IL-2, and IL-3, IL-4, and GM-CSF. IL-10 is also known to suppress antigen presentation on antigen presenting cells, enhances B cell survival, proliferation, and antibody production. IL-10 can block NF-kappa B activity, and is involved in the regulation of the JAK-STAT signaling pathway. While human IL-10 can act on murine cells, murine IL-10 cannot act on human cells. Binds to a retinoid X receptor (RXR) responsive element from the cellular retinol-binding protein II promoter (CRBPII-RXRE). IL-10 inhibits the 9-cis-retinoic acid-dependent RXR alpha transcription activation of the retinoic acid responsive element. Knockout studies in mice suggest the function of this cytokine as an essential immunoregulator in the intestinal tract.
Specifications

Specifications

P22301
protein with no preservative
18.6
E. coli
RUO
CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF
Human
Recombinant
Lyophilized
ELISA, Functional Assay, Western Blot
3586
Human IL-10
-20°C
<1 EU/μg
IL10
Unconjugated
MSPGQGTQSE NSCTHFPGNL PNMLRDLRDA FSRVKTFFQM KDQLDNLLLK ESLLEDFKGY LGCQALSEMI QFYLEEVMPQ AENQDPDIKA HVNSLGENLK TLRLRLRRCH RFLPCENKSK AVEQVKNAFN KLQEKGIYKA MSEFDIFINY IEAYMTMKIR N
≥ 98% by SDS-PAGE gel and HPLC analyses.
Videos
SDS
Documents

Documents

Product Certifications