Learn More
R&D Systems™ Human IL-17 Quantikine HS ELISA Kit
Human IL-17 Quantikine HS ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand:  R&D Systems™ HS170
Product Code. 18730877
  • 980.00 € / Pack of 96
Explore more special offers
Description
Description
The Human IL-17 Quantikine HS ELISA Kit quantifies human IL-17/IL-17A in serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
Specifications
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Human IL-17 Quantikine HS ELISA Kit
Microplate, conjugate, standards, assay diluents, calibrator diluents, wash buffer, color reagent, stop Solution, plate sealers
4 hours
Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
3605 (Human); 16171 (Mouse); 301289 (Rat); 481837 (Canine)
Measure IL-17 levels in serum and plasma