missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-8/CXCL8 Quantikine HS ELISA Kit

Product Code. 18751794
Click to view available options
Quantity:
1 Pack (6 Plates)
One 96-well plate
Unit Size:
1 set
Pack of 96
This item is not returnable. View return policy

Product Code. 18751794

Brand: R&D Systems™ HS800

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human IL-8/CXCL8 Quantikine HS ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-8/CXCL8 Quantikine HS ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CXCL8/IL-8 in serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 1.00 - 64 pg/mL
Assay Sensitivity 0.4 pg/mL
Conjugate HRP
Sample Type Plasma (EDTA), Heparin Plasma, Serum, Serum
Specificity Natural and Recombinant Human IL-8
For Use With (Application) ELISA
Gene ID (Entrez) 3576
Quantity One 96-well plate
Sample Volume Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym 3-10 C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1 TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1 NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10 C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.