Learn More
Invitrogen™ IL-8 Human Uncoated ELISA Kit with Plates
Specifically engineered for accurate and precise measurement of IL-8 protein levels from samples including serum, plasma, and cell culture supernatants. This second generation kit has increased sensitivity with a range from 2-250pg/mL.
Specifications
Accession Number | P10145 |
---|---|
Assay Range | 2-250 pg/mL |
Assay Sensitivity | 2 pg/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Description
The Human Interleukin 8 (IL-8) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IL-8. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Human IL-8 Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)Specifications
P10145 | |
2 pg/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
GCP-1,GCP1,IL8,LECT,LUCT,LYNAP,MDNCF,MONAP,NAF,NAP-1,NAP1 | |
HRP | |
Plasma, 100 μL; Serum, 100 μL; Supernatant, 100 μL | |
Interleukin-8, CXCL8, NAP-I | |
1 hr. 45 min. |
2-250 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
ELISA | |
3576 | |
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, Coating Buffer: 10X PBS ELISA Coating Buffer, Assay Diluent: 5X Concentrated, Detection Enzyme: Pre-titrated Avidin-HRP, Substrate Solution: Tetramethylbenzidine (TMB) Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards., 96-well Plates: Corning Costar flat-bottom plates (included with Cat. Nos. ending in -22 and -86) | |
RUO | |
2°C to 8°C | |
Human | |
24 hr. 30 min. |
For Research Use Only