missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human LOXL4 (NP_115587.6) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Specifications
Specifications
| Gene | LOXL4 |
| Accession Number | NP_115587.6 |
| Gene ID (Entrez) | 84171 |
| Gene Alias | EC 1.4.3, EC 1.4.3.-, FLJ21889, LOX L4, LOXC, LOXClysyl oxidase related C, lysyl oxidase homolog 4, lysyl oxidase-like 4, Lysyl oxidase-like protein 4, Lysyl oxidase-related protein C |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction