Learn More
Invitrogen™ Human LXR alpha (aa 318-329) Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP114
Product Code. 11514770
  • 190.00 € / 50µg
Explore more special offers
Description
Description
Human LXR alpha (residues 318-329) Synthetic Peptide, PEP-114, from Invitrogen.
PEP-114 is a 12 amino acid synthetic peptide whose sequence corresponds to residues 318-329 of human LXR alpha with an amino-terminal cysteine added for carrier protein conjugation. The sequence of this peptide is (amino to carboxy terminus): C-D318-F-S-Y-N-R-E-D-F-A-K-A (329).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human LXR alpha, catalog # PA1-330. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10062
purified
Human LXR alpha (aa 318-329)
-20°C. Avoid freeze-thaw cycles.
Lyophilized
Control
Yes
Peptide
RUO - research use only
Human
Q13133
0.5 mg/mL
Synthetic Peptide
peptide with no preservative
NR1H3
No preservative
50 μg
C-D318-F-S-Y-N-R-E-D-F-A-K-A(329)