Learn More
Invitrogen™ Human LXR alpha (aa 432-442) Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP115
Product Code. 11524770
  • 181.00 € / 50µg
Explore more special offers
Description
Description
Human LXR Alpha (residues 432-442) Synthetic Peptide, PEP-115, from Invitrogen.
PEP-115 is a 14 amino acid synthetic peptide whose sequence corresponds to residues 432-445 of human LXR alpha with an amino-terminal cysteine added for carrier protein conjugation. The sequence of this peptide is (amino to carboxy terminus): C-L432-R-L-Q-D-K-K-L-P-P-L-L-S-E445This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human LXR alpha and beta, catalog # PA1-331B. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10062
purified
Human LXR alpha (aa 432-442)
-20°C. Avoid freeze-thaw cycles.
Lyophilized
Control
Yes
Peptide
RUO - research use only
Human
Q13133
0.5 mg/mL
Synthetic Peptide
peptide with no preservative
NR1H3
No preservative
50 μg
C-L432-R-L-Q-D-K-K-L-P-P-L-L-S-E445