missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human M-CSF Recombinant Protein
M-CSF (macrophage colony-stimulating factor) is a bioactive protein intended for use in cell culture applications
256.00 € - 1460.00 €
Specifications
For Use With (Application) | Cell Culture |
---|---|
Molecular Weight (g/mol) | 38 kDa |
Gene Alias | M-CSF |
Biological Activity | ED50 = ≤ 5 ng⁄mL |
Species | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10089418
|
Gibco™
PHC9504 |
10 μg |
256.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10336479
|
Gibco™
PHC9501 |
100 μg |
1460.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Human M-CSF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
M-CSF induces proliferation and differentiation of hematopoietic stem cells into macrophages and growth and differentiation of monocytes.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: proteins bioassayed with Gibco media
- High-purity, recombinant, bioactive protein: to help ensure our growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High biological activity: in-house activity testing of our recombinant proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by measuring the dose dependent stimulation of proliferation of mouse M-NFS-60 cells
Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
M-CSF | |
Human | |
< 0.1 ng/μg | |
Mammalian | |
Recombinant, Ligand | |
GM-CSF, G-CSF, & M-CSF | |
Gibco™ |
38 kDa | |
ED50 = ≤ 5 ng⁄mL | |
Store this lyophilized preparation at 2-8°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at ≤ -20°C. Avoid repeated freeze⁄thaw cycles. | |
Wet Ice | |
Cytokines & Receptors | |
Lyophilized | |
Cardiovascular Research, Stem Cell Research, Differentiation, Inflammation | |
>95% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title