Learn More
R&D Systems™ Human MBP DuoSet ELISA
This DuoSet ELISA Development kit contains the basic components required for the development of sandwich ELISAs
Brand:  R&D Systems™ DY4228-05
Product Code. 30020967
  • 468.00 € / 1 set
Explore more special offers
Includes: Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Description
Description
The Human MBP DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human MBP.
Specifications
Specifications
1 Kit (for 5 Plates)
Biotin
234.40 to 15000 pg/mL
4155
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Human
MBP
ELISA
Hmbpr, MGC99675, mld, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein, shi