Learn More
Invitrogen™ Human MEP1B Control Fragment Recombinant Protein
Recombinant Protein
Brand:  Invitrogen™ RP102555
Product Code. 30199588
  • 259.00 € / 100µL
Explore more special offers
Description
Description
Highest antigen sequence indentity to the following orthologs: Mouse (68%), Rat (68%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83064 (PA5-83064. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Meprins are multidomain zinc metalloproteases that are highly expressed in mammalian kidney and intestinal brush border membranes, and in leukocytes and certain cancer cells. They are involved in the hydrolysis of a variety of peptide and protein substrates, and have been implicated in cancer and intestinal inflammation. Mature meprins are oligomers of evolutionarily related, but separately encoded alpha and/or beta subunits. Homooligomers of alpha subunit are secreted, whereas, oligomers containing the beta subunit are plasma membrane-bound. This gene encodes the beta subunit. Targeted disruption of this gene in mice affects embryonic viability, renal gene expression profiles, and distribution of the membrane-associated alpha subunit in kidney and intestine.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Q16820
Blocking Assay, Control
4225
100 μL
RUO
MEP1B
Unconjugated
Recombinant
MTTDNGNYFWDRPSKVGTVALFPNGTQFRRGGGYGTSAFITHERLKSRDFIKGDDVYILLTVEDISHLNSTQIQLTPAPSVQDLCSKTTCKNDGVCTVRDGKAECRC
E. coli
>80% by SDS-PAGE and Coomassie blue staining
≥5.0 mg/mL
1 M urea, PBS with no preservative; pH 7.4
Human MEP1B Control Fragment
-20°C, Avoid Freeze/Thaw Cycles
EC 3.4.24.63; Endopeptidase-2; MEP1B; Mep-1 b; meprin 1 beta; meprin A; meprin A subunit beta; meprin A, beta; meprin B; meprin beta; N-benzoyl-L-tyrosyl-p-amino-benzoic acid hydrolase beta subunit; N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit beta; PABA peptide hydrolase; PPH beta
Mep1b
Human
His-ABP-tag
-20°C, Avoid Freeze/Thaw Cycles