missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human/Mouse FGF-8b Recombinant Protein, PeproTech®
E. coli Recombinant Protein
80.00 € - 2965.00 €
Specifications
Accession Number | P37237, P55075 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 14179, 2253 |
Molecular Weight (g/mol) | 22.5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17881043
|
Gibco™
100-25-100UG |
100 μg |
594.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17870803
|
Gibco™
100-25-1MG |
2 x 500 μg |
2965.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17800293
|
Gibco™
100-25-250UG |
250 μg |
1040.00 €
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17881053
|
Gibco™
100-25-25UG |
25 μg |
194.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17880023
|
Gibco™
100-25-500UG |
500 μg |
1776.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17820013
|
Gibco™
100-25-5UG |
5 μg |
80.00 €
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGF-8 (FGF-8b) is a heparin-binding growth factor belonging to the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. There are 4 known alternate spliced forms of FGF8; FGF-8A, FGF-8B, FGF-8E and FGF-8F. The human and murine FGF-8A and B are identical, unlike human and mouse FGF-8E and F, which are 98% identical. FGF-8 targets mammary carcinoma cells and other cells expressing the FGF receptors.Specifications
P37237, P55075 | |
protein with no preservative | |
22.5 | |
E. coli | |
RUO | |
FGF8 | |
Unconjugated | |
MQVTVQSSPN FTQHVREQSL VTDQLSRRLI RTYQLYSRTS GKHVQVLANK RINAMAEDGD PFAKLIVETD TFGSRVRVRG AETGLYICMN KKGKLIAKSN GKGKDCVFTE IVLENNYTAL QNAKYEGWYM AFTRKGRPRK GSKTRQHQRE VHFMKRLPRG HHTTEQSLRF EFLNYPPFTR SLRGSQRTWA PEPR | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
14179, 2253 | |
Human/Mouse FGF-8 b | |
-20°C | |
<1 EU/μg | |
Human, Mouse | |
Recombinant | |
Lyophilized |