Learn More
Invitrogen™ Human MRP3 Vesicles
Human MRP3 Vesicles are insect derived purified plasma membranes with inserted MRP3 transport protein (Multidrug Resistance-associated Protein 3, ABCC3). Our Human MRP3 Vesicles can be used to evaluate test compounds and drug interactions with MRP3 in in vitro assays.
Brand: Invitrogen™ GM0021
Description

Specifications
Frozen | |
Research Grade | |
Human Vesicle | |
Vial |
0.5 mL | |
Human | |
Store at -80°C. | |
Dry Ice |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.