missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MUNC18 (aa 58-70) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP067
This item is not returnable.
View return policy
Description
Human Munc-18 (residues 58-70) Synthetic Peptide, PEP-067, from Invitrogen.
PEP-067 is a 13 amino acid synthetic phospho-peptide whose sequence corresponds to residues 58-70 of human Munc-18. The sequence of this peptide is (amino to carboxy terminus): V58-E-D-I-N-K-R-R-E-P-I-P-S70 ©This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and Munc-18, catalog # PA1-742. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
Specifications
6812 | |
purified | |
Human MUNC18 (aa 58-70) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
P61764 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
STXBP1 | |
No preservative | |
50 μg | |
V58 - E - D - I - N - K - R - R - E - P - I - P - S70 (C) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction