Learn More
Invitrogen™ Human Nectin-2/CD112 ELISA Kit
Human Nectin-2/CD112 quantitates human Nectin-2/CD112 in serum, plasma, supernatant.
Brand:  Invitrogen™ EH331RB
 View more versions of this product
Product Code. 17104633
  • 654.00 € / Pack of 96
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Human Nectin-2/CD112 quantitates human Nectin-2/CD112 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Nectin-2/CD112.
This gene encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions. It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AI325026; AI987993; Cd112; Herpes virus entry mediator B; herpesvirus entry mediator B; herpesvirus entry protein B; hveB; mHveB; Mph; Murine herpes virus entry protein B; murine herpesvirus entry protein B; nectin cell adhesion molecule 2; NECTIN2; Nectin-2; Poliovirus receptor homolog; poliovirus receptor related 2; poliovirus receptor-like 2; poliovirus receptor-related 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); poliovirus receptor-related protein 2; poliovirus sensitivity; PRR2; Pvr; Pvrl2; PVRR2; Pvs
0.05 ng/mL
ELISA
Colorimetric Microplate Reader
NECTIN2
<10%
HRP
Research Use Only
2°C to 8°C
1 hr 20 min
0.05 to 10 ng/mL
Biotin
Plasma, Serum, Supernatant
Q92692
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Supernatant, 50 μL; Serum, 50 μL; Plasma, 50 μL
Nectin 2
4 hr 45 min