missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Nectin-2/CD112 ELISA Kit
Human Nectin-2/CD112 quantitates human Nectin-2/CD112 in serum, plasma, supernatant.
654.00 € - 2635.00 €
Specifications
Accession Number | AI325026; AI987993; Cd112; Herpes virus entry mediator B; herpesvirus entry mediator B; herpesvirus entry protein B; hveB; mHveB; Mph; Murine herpes virus entry protein B; murine herpesvirus entry protein B; nectin cell adhesion molecule 2; NECTIN2; Nectin-2; Poliovirus receptor homolog; poliovirus receptor related 2; poliovirus receptor-like 2; poliovirus receptor-related 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); poliovirus receptor-related protein 2; poliovirus sensitivity; PRR2; Pvr; Pvrl2; PVRR2; Pvs |
---|---|
Assay Range | 0.05 to 10 ng/mL |
Assay Sensitivity | 0.05 ng/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17105109
|
Invitrogen™
EH331RBX5 |
5 x 96 Tests |
2635.00 €
Each |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||||
17104633
|
Invitrogen™
EH331RB |
96 Tests |
654.00 €
Pack of 96 |
Estimated Shipment: 03-01-2025
Log in to see stock. |
|||||
Description
Human Nectin-2/CD112 quantitates human Nectin-2/CD112 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Nectin-2/CD112.
This gene encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions. It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
AI325026; AI987993; Cd112; Herpes virus entry mediator B; herpesvirus entry mediator B; herpesvirus entry protein B; hveB; mHveB; Mph; Murine herpes virus entry protein B; murine herpesvirus entry protein B; nectin cell adhesion molecule 2; NECTIN2; Nectin-2; Poliovirus receptor homolog; poliovirus receptor related 2; poliovirus receptor-like 2; poliovirus receptor-related 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); poliovirus receptor-related protein 2; poliovirus sensitivity; PRR2; Pvr; Pvrl2; PVRR2; Pvs | |
0.05 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
NECTIN2 | |
<10% | |
HRP | |
Supernatant, 50 μL; Serum, 50 μL; Plasma, 50 μL | |
Nectin 2 | |
4 hr 45 min |
0.05 to 10 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Q92692 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |