Learn More
Invitrogen™ Human NOD2 Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP0306
Product Code. 15246519
  • 141.00 € / 50µg
Explore more special offers
Description
Description
NOD2 Synthetic Peptide, PEP-0306, from Invitrogen.
NOD2 is a member of the apoptosis regulating protein family that includes caspase recruitment-domains, as well as Apaf-1 and NOD1. It contains two N-terminal CARDs, a nucleotide binding domain (NBD), and multiple C-terminal leucine-rich repeats (LRRs). NOD2 is expressed in monocytes (whereas NOD1 is expressed in multiple tissues). NOD2 plays a role in regulating NF-kappaB, acts as an intracellular receptor for bacterial lipopolysaccharides, and contributes to inflammatory bowel disease (IBD) and Crohn's disease.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
64127
purified
Human NOD2
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
Q9HC29
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
NOD2
0.02% sodium azide
50 μg
PBS
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Human NOD2 Synthetic Peptide > 50 μg; Unlabeled