Learn More
Novus Biologicals™ Human Normal Tissue MicroArray (Normal)
Tissue arrays are 4 - 6 um in thickness, and mounted on positively charged glass slides
Brand:  Novus Biologicals™ NBP2-42042
Product Code. 18602038
  • 459.00 € / Pack of 2
Explore more special offers
Specifications
2 Slides
Human
Immunohistochemistry, Immunohistochemistry-Paraffin