Learn More
R&D Systems™ Human NOX1 (NP_008983) VersaClone cDNA
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
Brand:  R&D Systems™ RDC2683
Product Code. 18700389
  • 309.00 € / 10µg
Explore more special offers
Description
Description
  • Also known as: MOX1; NOH1; NOH-1; GP91-2
  • NOX1 is a member of the NADPH oxidase family of enzymes responsible for the catalytic oneelectron transfer of oxygen to generate superoxide or hydrogen peroxide.
  • It participates in the regulation of cellular pH and is blocked by zinc.
  • Alternatively spliced transcripts encoding different proteins have been described.
  • The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
  • For reasearch use only. Not for use in humans.
Specifications
Specifications
hNOX1bed
10 μg at 0.2 μg/μL
1708 bp
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
NP_008983
cDNA is provided in 10 mM Tris-Cl, pH 8.5
27035