missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human PDGF-AB Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 100-00AB-500UG
This item is not returnable.
View return policy
Description
Platelet-Derived Growth Factor (PDGF) is a mitogenic peptide growth hormone carried in the alpha-granules of platelets and is released when platelets adhere to traumatized tissues. Connective tissue cells near the traumatized region respond by initiating the process of replication. The synthesis of PDGF can be induced by IL-1, IL-6, TNF-alpha, TGF-beta and EGF.Specifications
P01127, P04085 | |
protein with no preservative | |
26.4 | |
500 μg | |
-20°C | |
<1 EU/μg | |
PDGFA, PDGFB | |
Unconjugated | |
Alpha chain: SIEEAVPAVC KTRTVIYEIP RSQVDPTSAN FLIWPPCVEV KRCTGCCNTS SVKCQPSRVH HRSVKVAKVE YVRKKPKLKE VQVRLEEHLE CACATTSLNP DYREEDTGRP RESGKKRKRK RLKPT Beta chain: SLGSLTIAEP AMIAECKTRT EVFEISRRLI DRTNANFLVW PPCVEVQRCS GCCNNRNVQC RPTQVQLRPV QVRKIEIVRK KPIFKKATVT LEDHLACKCE TVAAARPVT | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
5154, 5155 | |
Human PDGF-AB | |
E. coli | |
RUO | |
Becaplermin; c-sis; H-PDGF-AB; IBGC5; PDGF A-chain; PDGF subunit A; PDGF subunit B; PDGF, B chain; PDGF1; PDGF-1; PDGF2; PDGF-2; Pdgfa; PDGF-A; PDGFB; PDGF-B; platelet derived growth factor subunit A; platelet derived growth factor subunit B; platelet derived growth factor, alpha; platelet derived growth factor, B polypeptide; platelet-derived growth factor 2; platelet-derived growth factor A chain; platelet-derived growth factor A-chain; platelet-derived growth factor alpha chain; platelet-derived growth factor alpha polypeptide; Platelet-derived growth factor B chain; platelet-derived growth factor beta polypeptide; platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis) oncogene homolog); platelet-derived growth factor subunit A; Platelet-derived growth factor subunit B; platelet-derived growth factor, beta polypeptide (oncogene SIS); proto-oncogene c-Sis; SIS; SSV | |
Human | |
Recombinant | |
Lyophilized |