missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human POMC (aa 192-267) Control Fragment Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP106110
This item is not returnable.
View return policy
Description
Highest antigen sequence indentity to the following orthologs: Mouse (62%), Rat (62%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes a polypeptide hormone precursor that undergoes extensive, tissue-specific, post-translational processing via cleavage by subtilisin-like enzymes known as prohormone convertases. There are eight potential cleavage sites within the polypeptide precursor and, depending on tissue type and the available convertases, processing may yield as many as ten biologically active peptides involved in diverse cellular functions. The encoded protein is synthesized mainly in corticotroph cells of the anterior pituitary where four cleavage sites are used; adrenocorticotrophin, essential for normal steroidogenesis and the maintenance of normal adrenal weight, and lipotropin beta are the major end products. In other tissues, including the hypothalamus, placenta, and epithelium, all cleavage sites may be used, giving rise to peptides with roles in pain and energy homeostasis, melanocyte stimulation, and immune modulation. These include several distinct melanotropins, lipotropins, and endorphins that are contained within the adrenocorticotrophin and beta-lipotropin peptides. Mutations in this gene have been associated with early onset obesity, adrenal insufficiency, and red hair pigmentation. Alternatively spliced transcript variants encoding the same protein have been described.
Specifications
P01189 | |
Blocking Assay, Control | |
5443 | |
100 μL | |
ACTH; adrenal corticotropic hormone; adrenocorticotropic hormone; adrenocorticotropin; alpha-melanocyte stimulating hormone; alpha-melanocyte-stimulating hormone; alphaMSH; alpha-MSH; BE; beta-endorphin; Beta-LPH; beta-melanocyte-stimulating hormone; beta-MSH; CLIP; Corticotropin; Corticotropin-like intermediary peptide; corticotropin-lipotropin; Gamma-LPH; gamma-MSH; lipotr; Lipotropin beta; lipotropin gamma; LPH; Melanocyte-stimulating hormone alpha; Melanocyte-stimulating hormone beta; Melanotropin alpha; melanotropin beta; Melanotropin gamma; met-enkephalin; MSH; Npp; opiomelanocortin prepropeptide; OTTHUMP00000119991; OTTHUMP00000200964; POC; Pomc; Pomc1; Pomc-1; Pomc2; Potential peptide; Precursor of MSH; pro-ACTH-endorphin; proopiomelanocortin; pro-opiomelanocortin; proopiomelanocortin (adrenocorticotropin/ beta-lipotropin/ alpha-melanocyte stimulating hormone/ beta-melanocyte stimulating hormone/ beta-endorphin); proopiomelanocortin preproprotein; proopiomelanocortin, beta (endorphin, beta); pro-opiomelanocortin-alpha; proopoimelanocortin, beta (endorphin, beta) | |
POMC | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human POMC (aa 192-267) Control Fragment | |
RUO | |
POMC | |
Unconjugated | |
Recombinant | |
DGPADDGAGAQADLEHSLLVAAEKKDEGPYRMEHFRWGSPPKDKRYGGFMTSEKSQTPLVTLFKNAIIKNAYKKGE | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Human POMC (aa 192-267) Control Fragment Recombinant Protein > 100 μL; Unlabeled
Spot an opportunity for improvement?Share a Content Correction