Learn More
R&D Systems™ Human Proprotein Convertase 4 (NP_060043.2) VersaClone cDNA
Brand:  R&D Systems™ RDC3601
Product Code. 18724497
  • 719.00 € / 10µg
Explore more special offers
Specifications
Proprotein convertase PC4
54760
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
NP_060043.2
DKFZp434B217, EC 3.4.21, EC 3.4.21.75, EC 3.4.21.93, MGC34749, PC4, PC4EC 3.4.21.-, Proprotein convertase 4, proprotein convertase subtilisin/kexin type 4, SPC5