Learn More
Novus Biologicals™ Human Prostate Tissue Lysate (Adult Tumor)
Vials of lysate containing 0.1mg of normal or prostatic lysate that has not been subjected to denaturing or reducing conditions. Lysates are guaranteed for 6 months from date of receipt.
Brand:  Novus Biologicals™ NBP2-47065
Product Code. 18116269
  • 474.00 € / 0.10mg
Explore more special offers
Description
Description
These lysates have not been subjected to denaturing or reducing conditions. This allows the tissue or cell lysate to be used in a variety of applications; to study protein-protein interaction, ligand binding, ELISA, immunoprecipitation, 1D and 2D gel electrophoresis, and Western blotting for the detection of specific protein targets. For use in 1D and 2D gel electrophoresis, the addition of a denaturing gel loading buffer with reducing agents may be required. Buffer requirements for performing protein-protein interaction and ligand binding studies can vary significantly from RIPA buffer and may require modifications. In most cases, tissue lysates in RIPA buffer can be used, directly in standard ELISA and immunoprecipitation assays. These lysates are proteomic discovery tools. Researchers should validate and optimize for individual use.
The Human Prostate Tissue Lysate (Adult) is derived from human.
Specifications
Specifications
Protein-Protein Interaction, Ligand Binding, ELISA, Immunoprecipitation, 1 D and 2 D Gel Electrophoresis, Western Blot
0.1 mg
Store at -80 C. Avoid freeze-thaw cycles.
Prostate
Human
Prostate