missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human PTHrP Recombinant Protein, PeproTech®
E. coli Recombinant Protein
82.00 € - 1518.00 €
Specifications
Accession Number | P12272 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 5744 |
Molecular Weight (g/mol) | 9.8 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17840553
|
Gibco™
100-09-100UG |
100 μg |
304.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17820553
|
Gibco™
100-09-10UG |
10 μg |
82.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17840143
|
Gibco™
100-09-1MG |
1 mg |
1518.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17880473
|
Gibco™
100-09-250UG |
250 μg |
532.00 €
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17830143
|
Gibco™
100-09-500UG |
500 μg |
912.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17840293
|
Gibco™
100-09-50UG |
50 μg |
199.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a member of the parathyroid hormone family. This hormone regulates endochondral bone development and epithelial-mesenchymal interactions during the formation of the mammary glands and teeth. This hormone is involved in lactation possibly by regulating the mobilization and transfer of calcium to the milk. The receptor of this hormone, PTHR1, is responsible for most cases of humoral hypercalcemia of malignancy. Four alternatively spliced transcript variants encoding two distinct isoforms have been observed. There is also evidence for alternative translation initiation from non-AUG (CUG and GUG) start sites, in-frame and downstream of the initiator AUG codon, to give rise to nuclear forms of this hormone.Specifications
P12272 | |
protein with no preservative | |
9.8 | |
E. coli | |
RUO | |
BDE2; calciotropic hormone, growth factor; HHM; osteostatin; parathyroid hormone like hormone; parathyroid hormone related peptide; parathyroid hormone-like hormone; parathyroid hormone-like hormone preproprotein; parathyroid hormone-like peptide; parathyroid hormone-like protein; parathyroid hormone-like related protein; parathyroid hormone-related peptide; parathyroid hormone-related protein; parathyroid-like peptide; PLP; PTHLH; PTH-like; PTHR; PTH-related peptide; PTH-related protein; Pthrp; PTH-rP; PTHrP[107-139 ]; PTHrP[1-36 ]; PTHrP[38-94 ] | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
5744 | |
Human PTHrP | |
-20°C | |
<1 EU/μg | |
PTHLH | |
Unconjugated | |
AVSEHQLLHD KGKSIQDLRR RFFLHHLIAE IHTAEIRATS EVSPNSKPSP NTKNHPVRFG SDDEGRYLTQ ETNKVETYKE QPLKTP | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title