Learn More
Invitrogen™ Human RAR gamma-2 (aa 13-28) Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP229
Product Code. 11594870
  • 173.00 € / 50µg
Explore more special offers
Description
Description
Human RAR gamma 2 (residues 13-28) Synthetic Peptide, PEP-229, from Invitrogen.
PEP-229 is a 16 amino acid synthetic peptide corresponding to amino acid residues 13-28 from human retinoic acid receptor gamma 2. The sequence of this peptide is (amino to carboxy terminus): R (13) R L Y G A A G P G A G L L R R (28).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, catalog # PA3-814. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5916
purified
Human RAR gamma-2 (aa 13-28)
-20°C. Avoid freeze-thaw cycles.
Lyophilized
Control
Yes
Peptide
RUO - research use only
Human
P13631
0.5 mg/mL
Synthetic Peptide
peptide with no preservative
RARG
No preservative
50 μg
R(13) R L Y G A A G P G A G L L R R(28)